• USA Home
  • 32488 - Chloramphenicol-13C11 solution

EMAIL THIS PAGE TO A FRIEND
32488 Supelco

Chloramphenicol-13C11 solution

25 μg/mL in acetonitrile, VETRANAL, analytical standard

Synonym: D-threo-2,2-Dichloro-N-[β-hydroxyα-(hydroxymethyl)-β-(4-nitrophenyl)ethyl]acetamide-13C11 solution

  • Empirical Formula (Hill Notation) 13C11H12Cl2N2O5

  • Molecular Weight 334.05

  •  EC Number 200-835-2

Purchase

Properties

Related Categories All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Biotoxin and Mycotoxin Standards, Food and Cosmetic Component Standards,
grade   analytical standard
product line   VETRANAL
shelf life   limited shelf life, expiry date on the label
concentration   25 μg/mL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
format   single component solution
mass shift   M+11
storage temp.   −20°C

Description

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?