EMAIL THIS PAGE TO A FRIEND
32992 Supelco

Aspoxicillin

VETRANAL, analytical standard

  • CAS Number 63358-49-6

  • Empirical Formula (Hill Notation) C21H27N5O7S

  • Molecular Weight 493.53

  •  NACRES NA.76

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Chemical Structure,
grade   analytical standard
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
storage temp.   2-8°C

Description

General description

Aspoxicillin is classified under the β-lactam family of antibiotics.

Application

Aspoxicillin may be used as an analytical reference standard for the determination of the analyte in:
• Meat samples by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS).
• Fish, shrimp and eel by LC coupled to quadrupole high resolution MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MB7548000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?