EMAIL THIS PAGE TO A FRIEND
33423 Sigma-Aldrich

1,2-Dibromoethane

PESTANAL®, analytical standard

Synonym: EDB, Ethylene dibromide

  • CAS Number 106-93-4

  • Linear Formula BrCH2CH2Br

  • Molecular Weight 187.86

  •  Beilstein Registry Number 605266

  •  EC Number 203-444-5

  •  MDL number MFCD00000233

  •  PubChem Substance ID 24860068

Purchase

Properties

grade   PESTANAL®, analytical standard
vapor density   ~6.5 (vs air)
vapor pressure   11.7 mmHg ( 25 °C)
InChI Key   PAAZPARNPHGIKF-UHFFFAOYSA-N
refractive index   n20/D 1.539(lit.)
bp   131-132 °C(lit.)
mp   8-11 °C (lit.)
density   2.18 g/mL at 25 °C(lit.)
Featured Industry   Agriculture
Environmental

Description

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1605 6.1 / PGI
WGK Germany 
3
RTECS 
KH9275000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

31040 1,2-Dibromoethane, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?