• USA Home
  • 33427 - 1,4:3,6-Dianhydro-D-mannitol

EMAIL THIS PAGE TO A FRIEND
33427 Sigma-Aldrich

1,4:3,6-Dianhydro-D-mannitol

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: Isomannide

  • CAS Number 641-74-7

  • Empirical Formula (Hill Notation) C6H10O4

  • Molecular Weight 146.14

  •  Beilstein/REAXYS Number 80509

  •  EC Number 211-374-1

  •  MDL number MFCD10698761

  •  PubChem Substance ID 24860075

Purchase

Properties

Quality Level   100
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D +89±2°, c = 5% in H2O
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   80-85 °C (lit.)
  82-87 °C
SMILES string   O[C@@H]1CO[C@@H]2[C@H](O)CO[C@H]12
InChI   1S/C6H10O4/c7-3-1-9-6-4(8)2-10-5(3)6/h3-8H,1-2H2/t3-,4-,5-,6-/m1/s1
InChI key   KLDXJTOLSGUMSJ-KVTDHHQDSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Preparation of monomers used in polymer chemistry; Building block

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?