EMAIL THIS PAGE TO A FRIEND
34061 Supelco

Furaltadon-d5

VETRANAL, analytical standard

Synonym: 5-[Morpholino(methyl-d2)]-3-(5-nitrofurfurylidenamino)-2-oxazolidinone-d3

  • CAS Number 1015855-64-7

  • Empirical Formula (Hill Notation) C13H11D5N4O6

  • Molecular Weight 329.32

  •  MDL number MFCD04973441

  •  PubChem Substance ID 329754786

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Chemical Class,
Quality Level   100
grade   analytical standard
InChI Key   YVQVOQKFMFRVGR-JDUWRFKESA-N
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
mass shift   M+5

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?