EMAIL THIS PAGE TO A FRIEND
341150 Sigma-Aldrich

Thionin perchlorate

95%

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes, T-U-V More...
assay   95%
mp   >300 °C (lit.)
λmax   600 nm
SMILES string   [O-]Cl(=O)(=O)=O.Nc1ccc2nc3ccc(N)cc3[s+]c2c1
InChI   1S/C12H10N3S.ClHO4/c13-7-1-3-9-11(5-7)16-12-6-8(14)2-4-10(12)15-9;2-1(3,4)5/h1-6H,13-14H2;(H,2,3,4,5)/q+1;/p-1
InChI key   ZXCLWPMKLDYDDF-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?