• USA Home
  • 34785 - 2,4-Di-tert-butylphenol

EMAIL THIS PAGE TO A FRIEND
34785 Sigma-Aldrich

2,4-Di-tert-butylphenol

purum, antioxidant, ≥98.0% (GC)

  • CAS Number 96-76-4

  • Linear Formula [(CH3)3C]2C6H3OH

  • Molecular Weight 206.32

  •  Beilstein/REAXYS Number 1910383

  •  EC Number 202-532-0

  •  MDL number MFCD00008828

  •  PubChem Substance ID 24861534

  •  NACRES NA.22

Purchase

Properties

vapor pressure   1 mmHg ( 84.5 °C)
grade   purum
assay   ≥98.0% (GC)
bp   265 °C (lit.)
mp   53-56 °C (lit.)
  54-58 °C
SMILES string   CC(C)(C)c1ccc(O)c(c1)C(C)(C)C
InChI   1S/C14H22O/c1-13(2,3)10-7-8-12(15)11(9-10)14(4,5)6/h7-9,15H,1-6H3
InChI key   ICKWICRCANNIBI-UHFFFAOYSA-N
Gene Information   human ... GABRA1(2554)

Description

Caution

may discolor to yellow on storage

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 2430 8 / PGIII
WGK Germany 
WGK 2
RTECS 
SK8260000
Flash Point(F) 
264.2 °F
Flash Point(C) 
129 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?