• USA Home
  • 35367 - Nitroxynil-(phenyl13C6)

EMAIL THIS PAGE TO A FRIEND
35367 Supelco

Nitroxynil-(phenyl13C6)

VETRANAL, analytical standard

Synonym: 4-Hydroxy-3-iodo-5-nitro-benzonitrile-13C6

  • CAS Number 1325559-31-6

  • Empirical Formula (Hill Notation) 13C6CH3IN2O3

  • Molecular Weight 295.97

  •  MDL number MFCD20036279

  •  PubChem Substance ID 329755135

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Pharmaceuticals, Illicit Drugs & Alcohol
Quality Level   100
grade   analytical standard
InChI Key   SGKGVABHDAQAJO-ZRDHQLPYSA-N
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
mass shift   M+6

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?