• USA Home
  • 35383 - Sulfadimethoxine-(phenyl-13C6)

EMAIL THIS PAGE TO A FRIEND
35383 Supelco

Sulfadimethoxine-(phenyl-13C6)

VETRANAL, analytical standard

Synonym: 4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzene-13C6-sulfonamide

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Chromatography,
grade   analytical standard
InChI Key   ZZORFUFYDOWNEF-CICUYXHZSA-N
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
mass shift   M+6

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?