• USA Home
  • 35388 - Toltrazuril-(N-methyl-d3)

EMAIL THIS PAGE TO A FRIEND
35388 Supelco

Toltrazuril-(N-methyl-d3)

VETRANAL, analytical standard

Synonym: 1-Methyl-d3-3-{3-methyl-4-[4-(trifluoromethylthio)phenoxy]phenyl}-1,3,5-triazine-2,4,6(1H,3H,5H)-trione

  • CAS Number 1353867-75-0

  • Empirical Formula (Hill Notation) C18D3H11F3N3O4S

  • Molecular Weight 428.40

  •  MDL number MFCD20264916

  •  PubChem Substance ID 329755150

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Pharmaceuticals, Illicit Drugs & Alcohol
Quality Level   100
grade   analytical standard
InChI Key   OCINXEZVIIVXFU-BMSJAHLVSA-N
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
mass shift   M+3

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?