EMAIL THIS PAGE TO A FRIEND
35598 Supelco

Gliotoxin solution

100 μg/mL in acetonitrile, analytical standard

Synonym: (3R,5aS,6S,10aR)-2,3,5a,6-Tetrahydro-6-hydroxy-3-(hydroxymethyl)-2-methyl-10H-3,10a-epidithiopyrazino[1,2-a]indole-1,4-dione, Aspergillin solution

Purchase

Properties

Related Categories All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Biotoxin and Mycotoxin Standards, Food and Cosmetic Component Standards,
grade   analytical standard
InChI Key   FIVPIPIDMRVLAY-RBJBARPLSA-N
shelf life   limited shelf life, expiry date on the label
concentration   100 μg/mL in acetonitrile
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
format   single component solution
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?