• USA Home
  • 358827 - 4′,5′-Dinitrofluorescein

EMAIL THIS PAGE TO A FRIEND
358827 Sigma-Aldrich

4′,5′-Dinitrofluorescein

Dye content 85 %

  • CAS Number 24545-86-6

  • Empirical Formula (Hill Notation) C20H10N2O9

  • Molecular Weight 422.30

  •  Beilstein/REAXYS Number 68725

  •  EC Number 246-308-0

  •  MDL number MFCD00042746

Purchase

Properties

composition   Dye content, 85%
mp   185 °C (dec.) (lit.)
SMILES string   Oc1ccc2c(Oc3c(ccc(O)c3[N+]([O-])=O)C24OC(=O)c5ccccc45)c1[N+]([O-])=O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?