• USA Home
  • 36065 - 2,4-Dichloro-6-nitrophenol

EMAIL THIS PAGE TO A FRIEND
36065 Sigma-Aldrich

2,4-Dichloro-6-nitrophenol

purum, ≥98.0% (HPLC)

  • CAS Number 609-89-2

  • Linear Formula Cl2C6H2(NO2)OH

  • Molecular Weight 208.00

  •  Beilstein Registry Number 2050081

  •  EC Number 210-202-2

  •  MDL number MFCD00007101

  •  PubChem Substance ID 329755269

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   LYPMXMBQPXPNIQ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   118-120 °C(lit.)
  121-124 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
SL0350000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?