• USA Home
  • 36523 - 4-Bromo-3-chloroaniline

EMAIL THIS PAGE TO A FRIEND
36523 Sigma-Aldrich

4-Bromo-3-chloroaniline

PESTANAL®, analytical standard

  • CAS Number 21402-26-6

  • Empirical Formula (Hill Notation) C6H5BrClN

  • Molecular Weight 206.47

  •  Beilstein Registry Number 1210160

  •  EC Number 244-370-3

  •  MDL number MFCD00041311

  •  PubChem Substance ID 329755374

Purchase

Properties

Related Categories A-B, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Pesticides & Metabolites Standards More...
grade   analytical standard
InChI Key   QLYHPNUFNZJXOQ-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?