• USA Home
  • 36679 - (S)-1-[(Rp)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine

EMAIL THIS PAGE TO A FRIEND
36679 Sigma-Aldrich

(S)-1-[(Rp)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine

≥97.0% (CHN/NMR)

Synonym: (1S)-1-(Dicyclohexylphosphino)-2-[(1S)-1-(diphenylphosphino)ethyl]ferrocene (acc to CAS), Josiphos SL-J004-2

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Other Chiral Ligands
InChI Key   AWOOIDCKLIQAJV-WLOLSGMKSA-N
assay   ≥97.0% (CHN/NMR)
optical activity   [α]20/D +99±5°, c = 0.4% in chloroform
application(s)   HPLC: suitable
  gas chromatography (GC): suitable

Description

Legal Information

sold in collaboration with Solvias AG

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?