• USA Home
  • 368644 - Oxalic bis(benzylidenehydrazide)

EMAIL THIS PAGE TO A FRIEND
368644 Sigma-Aldrich

Oxalic bis(benzylidenehydrazide)

98%

Purchase

Properties

assay   98%
mp   >300 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?