EMAIL THIS PAGE TO A FRIEND
36916 Supelco

4-Nitroanisole

analytical standard

Synonym: 1-Methoxy-4-nitrobenzene

  • CAS Number 100-17-4

  • Linear Formula O2NC6H4OCH3

  • Molecular Weight 153.14

  •  Beilstein/REAXYS Number 1865361

  •  EC Number 202-825-3

  •  MDL number MFCD00007327

  •  PubChem Substance ID 329755536

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
Quality Level   100
grade   analytical standard
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
density   1.233 g/mL at 25 °C (lit.)
Featured Industry   Environmental
format   neat
SMILES string   COc1ccc(cc1)[N+]([O-])=O
InChI   1S/C7H7NO3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3
InChI key   BNUHAJGCKIQFGE-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3458 6.1 / PGIII
WGK Germany 
WGK 1
RTECS 
BZ8800000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?