EMAIL THIS PAGE TO A FRIEND
37025 Supelco

Retrorsine

analytical standard

Synonym: β-Longilobine, 12,18-Dihydroxysenecionan-11,16-dione

  • CAS Number 480-54-6

  • Empirical Formula (Hill Notation) C18H25NO6

  • Molecular Weight 351.39

Purchase

Properties

Related Categories All alphabetically sorted, Analytical Standards, Analytical/Chromatography, Biotoxin and Mycotoxin Standards, Dried Down RMs,
grade   analytical standard
InChI Key   BCJMNZRQJAVDLD-CQRYIUNCSA-N
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   208-211 °C (lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
format   neat
storage temp.   −20°C

Description

Reconstitution

Dried down, 50μg/mL after reconstitution with 1 mL of water

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGII
WGK Germany 
3
RTECS 
VH7525000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?