• USA Home
  • 37420 - 1,3-Dihydroxyacetone dimer

EMAIL THIS PAGE TO A FRIEND
37420 Sigma-Aldrich

1,3-Dihydroxyacetone dimer

puriss.

Synonym: 2,5-Dihydroxydioxane-2,5-dimethanol

  • CAS Number 26776-70-5

  • Empirical Formula (Hill Notation) C6H12O6

  • Molecular Weight 180.16

  •  Beilstein/REAXYS Number 112910

  •  MDL number MFCD00051019

  •  PubChem Substance ID 24863288

Purchase

Properties

grade   puriss.
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
ign. residue   ≤1%
mp   75-80 °C (lit.)
storage temp.   2-8°C
SMILES string   OCC1(O)COC(O)(CO)CO1
InChI   1S/C6H12O6/c7-1-5(9)3-12-6(10,2-8)4-11-5/h7-10H,1-4H2
InChI key   KEQUNHIAUQQPAC-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D107204 1,3-Dihydroxyacetone dimer, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?