• USA Home
  • 37570 - 2,6-Dihydroxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
37570 Sigma-Aldrich

2,6-Dihydroxybenzoic acid

technical, ≥95% (T)

Synonym: γ-Resorcylic acid

  • CAS Number 303-07-1

  • Linear Formula (HO)2C6H3CO2H

  • Molecular Weight 154.12

  •  Beilstein/REAXYS Number 2209755

  •  EC Number 206-134-8

  •  MDL number MFCD00002462

Purchase

Properties

grade   technical
assay   ≥95% (T)
mp   165 °C (dec.) (lit.)
SMILES string   OC(=O)c1c(O)cccc1O
InChI   1S/C7H6O4/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3,8-9H,(H,10,11)
InChI key   AKEUNCKRJATALU-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DG8578000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D109606 2,6-Dihydroxybenzoic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?