• USA Home
  • 37738 - 1,6-Dihydroxynaphthalene

EMAIL THIS PAGE TO A FRIEND
37738 Sigma-Aldrich

1,6-Dihydroxynaphthalene

purum, ≥97.0% (HPLC)

Synonym: 1,6-Naphthalenediol

  • CAS Number 575-44-0

  • Linear Formula C10H6(OH)2

  • Molecular Weight 160.17

  •  Beilstein/REAXYS Number 1939032

  •  EC Number 209-386-7

  •  MDL number MFCD00003981

  •  PubChem Substance ID 24863499

Purchase

Properties

grade   purum
assay   ≥97.0% (HPLC)
mp   130-133 °C (lit.)
  136-140 °C
SMILES string   Oc1ccc2c(O)cccc2c1
InChI   1S/C10H8O2/c11-8-4-5-9-7(6-8)2-1-3-10(9)12/h1-6,11-12H
InChI key   FZZQNEVOYIYFPF-UHFFFAOYSA-N

Description

Caution

may discolor to gray or brown on storage

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
QJ4743333

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?