EMAIL THIS PAGE TO A FRIEND
37872 Supelco

Bentranil

PESTANAL®, analytical standard

Synonym: 2-Phenyl-3,1-benzoxazin-4-one

  • CAS Number 1022-46-4

  • Empirical Formula (Hill Notation) C14H9NO2

  • Molecular Weight 223.23

  •  Beilstein/REAXYS Number 170624

  •  MDL number MFCD00043598

  •  PubChem Substance ID 329755700

Purchase

Properties

Related Categories A-B, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Pesticides & Metabolites Standards More...
Quality Level   100
grade   analytical standard
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   189-192 °C/8 mmHg (lit.)
mp   123-125 °C (lit.)
Featured Industry   Agriculture
Environmental
format   neat
storage temp.   room temp
SMILES string   O=C1OC(=Nc2ccccc12)c3ccccc3
InChI   1S/C14H9NO2/c16-14-11-8-4-5-9-12(11)15-13(17-14)10-6-2-1-3-7-10/h1-9H
InChI key   HTTLBYITFHMYFK-UHFFFAOYSA-N

Description

General description

Bentranil is a herbicide, which is used for the postemergence control of annual weeds in cereal crops, rice and maize.

Application

Bentranil may be used as an analytical reference standard for the quantification of the analyte in roots of Murdannia lanuginosa and Murdannia simplex using gas chromatography coupled to mass spectrometry.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?