• USA Home
  • 379727 - Potassium oxalate monohydrate

EMAIL THIS PAGE TO A FRIEND
379727 Sigma-Aldrich

Potassium oxalate monohydrate

99.98% trace metals basis

Synonym: Ethanedioic acid, Oxalic acid potassium salt

  • CAS Number 6487-48-5

  • Linear Formula (COOK)2 · H2O

  • Molecular Weight 184.23

  •  Beilstein/REAXYS Number 3752576

  •  EC Number 209-506-8

  •  MDL number MFCD00150033

  •  PubChem Substance ID 57649785

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Potassium Salts, Research Essentials,
assay   99.98% trace metals basis
SMILES string   O.[K+].[K+].[O-]C(=O)C([O-])=O
InChI   1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2
InChI key   QCPTVXCMROGZOL-UHFFFAOYSA-L

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?