EMAIL THIS PAGE TO A FRIEND
38688 Supelco

Dodecanenitrile

analytical standard

Synonym: Lauronitrile, Undecyl cyanide

Purchase

Properties

grade   analytical standard
assay   ≥99.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤0.5% water
refractive index   n20/D 1.435-1.437
  n20/D 1.436 (lit.)
bp   198 °C/100 mmHg (lit.)
density   0.827 g/mL at 25 °C (lit.)
Featured Industry   Cleaning Products
Cosmetics
Food and Beverages
Personal Care
format   neat
SMILES string   CCCCCCCCCCCC#N
InChI   1S/C12H23N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-11H2,1H3
InChI key   VXCUURYYWGCLIH-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3276 6.1 / PGIII
WGK Germany 
2
RTECS 
JR2600000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?