EMAIL THIS PAGE TO A FRIEND
390127 Sigma-Aldrich

Activated charcoal Green Alternative

purified by resistive heating of graphite

Synonym: Charcoal activated

Purchase

Properties

Related Categories Activated and Mesoporous Carbon, Alternative Energy, Chemical Synthesis, Enabling Products, Greener Alternative Products,
vapor pressure   <0.1 mmHg ( 20 °C)
InChI Key   OKTJSMMVPCPJKN-UHFFFAOYSA-N
form   powder
autoignition temp.   842 °F
purified by   resistive heating of graphite
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
resistivity   1375 μΩ-cm, 20°C (graphite)
mp   3550 °C(lit.)

Description

Analysis Note

Fullerene yield 5-10%

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FF5800000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?