EMAIL THIS PAGE TO A FRIEND
39520 Sigma-Aldrich

2,6-Dimethylaniline

purum, ≥98.0% (GC)

Synonym: 2,6-Xylidine, 2-Amino-1,3-dimethylbenzene, 2-Amino-m-xylene

  • Linear Formula (CH3)2C6H3NH2

  • Molecular Weight 121.18

  •  Beilstein/REAXYS Number 636332

  •  EC Number 201-758-7

  •  MDL number MFCD00007747

  •  PubChem Substance ID 329755878

Purchase

Properties

vapor pressure   <0.01 mmHg ( 20 °C)
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.560 (lit.)
  n20/D 1.560
bp   214 °C/739 mmHg (lit.)
mp   10-12 °C (lit.)
density   0.984 g/mL at 25 °C (lit.)
SMILES string   Cc1cccc(C)c1N
InChI   1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3
InChI key   UFFBMTHBGFGIHF-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
RIDADR 
UN 1711 6.1 / PGII
WGK Germany 
2
RTECS 
ZE9275000
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?