• USA Home
  • 39580 - 5,6-Dimethylbenzimidazole

EMAIL THIS PAGE TO A FRIEND
39580 Sigma-Aldrich

5,6-Dimethylbenzimidazole

purum, ≥99.0% (NT)

  • CAS Number 582-60-5

  • Empirical Formula (Hill Notation) C9H10N2

  • Molecular Weight 146.19

  •  Beilstein Registry Number 116595

  •  EC Number 209-488-1

  •  MDL number MFCD00005603

  •  PubChem Substance ID 329755881

Purchase

Properties

Related Categories Benzimidazoles, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
InChI Key   LJUQGASMPRMWIW-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (NT)
mp   202-205 °C(lit.)
  202-205 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DD8280000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D147206 5,6-Dimethylbenzimidazole, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?