• USA Home
  • 39778 - Lidocaine-(diethyl-d10)

EMAIL THIS PAGE TO A FRIEND
39778 Supelco

Lidocaine-(diethyl-d10)

analytical standard

Synonym: 2-[Di(ethyl-d5)amino]-N-(2,6-dimethylphenyl)acetamide, [2H10]-Lidocaine

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Anesthetics Standards, Chromatography,
grade   analytical standard
InChI Key   NNJVILVZKWQKPM-JKSUIMTKSA-N
assay   ≥98.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
storage temp.   2-8°C

Description

Analysis Note

LC-MS identity conforms

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?