• USA Home
  • 39964 - Tryptone water, Vegitone

EMAIL THIS PAGE TO A FRIEND
39964 Sigma-Aldrich

Tryptone water, Vegitone

for microbiology

Synonym: Peptone water tryptic digest, Vegitone, Vegitone Tryptone water

Purchase

Description

Application

Vegitone Tryptone Water contains plant peptone instead of animal derived peptone. It is used for the detection of indole production by coliforms. It is also recommended by ISO Committee under the specification ISO 9308-1:1990.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

EZ-Product Family for Bioburden Testing

The EZ family has been designed to optimize and streamline your bioburden workflow. Ideal for microbiological water, beverage, and medical device testing using the membrane filtration method. The fam...
Keywords: Filtration

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?