• USA Home
  • 40220 - 4,4′-Dimethyl-2,2′-dipyridyl

EMAIL THIS PAGE TO A FRIEND
40220 Supelco

4,4′-Dimethyl-2,2′-dipyridyl

for spectrophotometric det., ≥99.0%

Synonym: 2,2′-Bi(γ-picoline), 4,4′-Dimethyl-2,2′-bipyridine

  • CAS Number 1134-35-6

  • Empirical Formula (Hill Notation) C12H12N2

  • Molecular Weight 184.24

  •  Beilstein/REAXYS Number 128995

  •  EC Number 214-483-2

  •  MDL number MFCD00006441

  •  PubChem Substance ID 329755955

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Spectroscopy, UV/Vis Reagents, UV/Visible (UV/VIS) Spectroscopy More...
grade   for spectrophotometric det.
InChI Key   NBPGPQJFYXNFKN-UHFFFAOYSA-N
assay   ≥99.0% (NT)
  ≥99.0%
mp   169-174 °C (lit.)
  171-173 °C

Description

General description

4,4′-Dimethyl-2,2′-dipyridyl is a monondentate ligand.

Application

It was used as a ligand in a study done to understand the photosubstitution in Tris chelate complexes of Ruthenium (II).

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DW1765000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?