EMAIL THIS PAGE TO A FRIEND
40594 Supelco

8-Geranyloxypsoralen

analytical standard

Synonym: 9-[[(2E)-3,7-Dimethyl-2,6-octadien-1-yl]oxy]-7H-furo[3,2-g][1]benzopyran-7-one

  • CAS Number 7437-55-0

  • Empirical Formula (Hill Notation) C21H22O4

  • Molecular Weight 338.40

  •  Beilstein Registry Number 313632

  •  MDL number MFCD03412052

  •  PubChem Substance ID 329756005

Purchase

Properties

Related Categories Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, Angelica, Citrus,
grade   analytical standard
InChI Key   SOVNCTNQAWWYAQ-OQLLNIDSSA-N
assay   ≥98.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤5.0% water
Featured Industry   Food and Beverages
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?