• USA Home
  • 407100 - 1,4-Dihydroxy-2,3-naphthalenedicarbonitrile

EMAIL THIS PAGE TO A FRIEND
407100 Sigma-Aldrich

1,4-Dihydroxy-2,3-naphthalenedicarbonitrile

97%

  • CAS Number 1018-79-7

  • Linear Formula (HO)2C10H4(CN)2

  • Molecular Weight 210.19

  •  Beilstein Registry Number 2695423

  •  MDL number MFCD00192049

  •  PubChem Substance ID 24865425

Purchase

Properties

Related Categories Cell Biology, D, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
InChI Key   JJBQXTABLSZHGC-UHFFFAOYSA-N
assay   97%
mp   >300 °C (lit.)

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Milli-Q® Water Purification Solutions
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?