• USA Home
  • 40915 - Silica gel 60 C8-reversed phase perfluorinated

EMAIL THIS PAGE TO A FRIEND
40915 Supelco

Silica gel 60 C8-reversed phase perfluorinated

Synonym: Perfluorinated silica gel 60

  •  NACRES SB.52

Purchase

Properties

Related Categories Chemical Synthesis, Fluorous Reversed-Phase Silica Gel, Fluorous Synthesis, Specialty Synthesis
Quality Level   100
form   powder
application(s)   LPLC: suitable
matrix active group   silica
particle size   0.035-0.070 mm
pore size   60 Å mean pore size
surface area   500 m2/g (approx.)

Description

General description

Reversed-phase silica gels are generally used to retain catalysts by hydrophobic interactions. Fluorous reversed phase silica gel (FRPSG) serves as an alternate for separation of perfluro-tagged compounds by solid phase extraction. FRPSG was used for the separation of steroids, flavonoids, polyphenols and taxanes. Purification of organic mixture was done using fluorous solid phase extraction (FSPE) on perfluorinated silica gel.

Application

Silica gel 60 C8-reversed phase perfluorinated was used in the purification of tertiary sulphonamide using FSPE.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?