• USA Home
  • 41162 - 4-Methoxy-2-methyl-2-butanethiol

EMAIL THIS PAGE TO A FRIEND
41162 Supelco

4-Methoxy-2-methyl-2-butanethiol

analytical standard

Synonym: 2-Mercapto-4-methoxy-2-methylbutane

  • CAS Number 94087-83-9

  • Empirical Formula (Hill Notation) C6H14OS

  • Molecular Weight 134.24

  •  Beilstein Registry Number 15016270

  •  EC Number 301-977-9

  •  MDL number MFCD08437076

  •  PubChem Substance ID 329756065

Purchase

Properties

Related Categories Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, Currant,
grade   analytical standard
InChI Key   XVHGKKGBUDMTIQ-UHFFFAOYSA-N
assay   ≥98.0% (GC)
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.451
density   0.915 g/mL at 20 °C
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
73.4 °F
Flash Point(C) 
23 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?