EMAIL THIS PAGE TO A FRIEND
41231 Supelco

Pentachlorobenzene

certified reference material, TraceCERT®

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, M-P, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   certified reference material
InChI Key   CEOCDNVZRAIOQZ-UHFFFAOYSA-N
grade   TraceCERT®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   275-277 °C(lit.)
mp   84-87 °C (lit.)
density   1.609 g/mL at 25 °C(lit.)
Featured Industry   Environmental
format   neat
storage temp.   2-8°C

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1325 - class 4.1 - PG 2 - RQ - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?