• USA Home
  • 41905 - Heptadecanoic acid tryptamide

EMAIL THIS PAGE TO A FRIEND
41905 Supelco

Heptadecanoic acid tryptamide

analytical standard

Synonym: MAT, Margaric acid tryptamide, N-Heptadecanoyltryptamine, N-[2-(3-Indolyl)ethyl]heptadecanamide

Purchase

Properties

Related Categories Additional Standards, Amides, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   ZVUYYSZDKRSPES-UHFFFAOYSA-N
assay   ≥99.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   112-116 °C
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?