EMAIL THIS PAGE TO A FRIEND
420433 Sigma-Aldrich

Talc

−350 mesh

Synonym: Hydrous magnesium silicate

Purchase

Properties

InChI Key   FPAFDBFIGPHWGO-UHFFFAOYSA-N
particle size   −350 mesh

Description

Packaging

2.5 kg in poly drum

500 g in poly bottle

Application

Talc may be used in the following processes:
• Preparation of polylactide (PLA)/talc composites by melt extrusion technique.
• Carrier in the dry inoculation method to get a dry inocula of Salmonella and Enterococcus faecium in low-moisture foods.
• Agent to create pleural adhesions.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
WW2710000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?