• USA Home
  • 42225 - Indium Standard for AAS

EMAIL THIS PAGE TO A FRIEND
42225 Supelco

Indium Standard for AAS

TraceCERT®, 1000 mg/L In in nitric acid

  •  NACRES NA.24

Purchase

Properties

Related Categories AAS, AAS CRMs, Additional Standards, Analytical Standards, Analytical/Chromatography,
Quality Level   300
grade   certified reference material
  TraceCERT®
concentration   1000 mg/L In in nitric acid
application(s)   AAS: suitable
format   single component solution

Description

General description

Indium is obtained as a byproduct of zinc processing. It can form alloys with a variety of precious and base metals.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the label.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Indium standard stock solution can be used in the determination of trace amount of indium in water samples, using graphite furnace atomic absorption spectrometry (GFAAS).

Analysis Note

1000 mg/L In in 2% nitric acid, prepared with high purity In metal, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?