• USA Home
  • 43366 - 4,5-Diphenyl-1,3-dioxol-2-one

EMAIL THIS PAGE TO A FRIEND
43366 Sigma-Aldrich

4,5-Diphenyl-1,3-dioxol-2-one

purum, ≥98.0% (HPLC)

Synonym: 1,2-Diphenylvinylene carbonate

  • CAS Number 21240-34-6

  • Empirical Formula (Hill Notation) C15H10O3

  • Molecular Weight 238.24

  •  Beilstein Registry Number 1114120

  •  EC Number 244-287-2

  •  MDL number MFCD00005381

Purchase

Properties

InChI Key   SROHGOJDCAODGI-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   71-75 °C(lit.)
  71-75 °C

Description

Other Notes

Protecting group reagent for prim. amines

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?