• USA Home
  • 434353 - 6,8-Dibromo-1′,3′-dihydro-1′,3′,3′-trimethylspiro[2H-1-benzopyran-2,2′-(2H)-indole]

EMAIL THIS PAGE TO A FRIEND
434353 Sigma-Aldrich

6,8-Dibromo-1′,3′-dihydro-1′,3′,3′-trimethylspiro[2H-1-benzopyran-2,2′-(2H)-indole]

97%

Synonym: SP-12

  • CAS Number 20200-62-8

  • Empirical Formula (Hill Notation) C19H17Br2NO

  • Molecular Weight 435.15

  •  MDL number MFCD00216613

  •  PubChem Substance ID 24867191

Purchase

Properties

assay   97%
mp   118-120 °C (lit.)

Description

Application

Photochromic dye

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?