EMAIL THIS PAGE TO A FRIEND
43592 Millipore

Tryptic Soy Broth

for microbiology, ready to use media in the bottle

Synonym: CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB, Tryptone Soya Broth

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), Bacillus, Bacteria (General Media), Bacteroides,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   casein peptone (pancreatic), 17 g/L
  dipotassium hydrogen phosphate, 2.5 g/L
  glucose, 2.5 g/L
  sodium chloride, 5 g/L
  soya peptone (papain digest.), 3 g/L
final pH   7.3±0.2 (25 °C)
Featured Industry   Clinical Testing
Cosmetics, Personal Care, and Cleaning Products
Environmental
Food and Beverages
Veterinary
suitability   nonselective for Bacillus
  nonselective for Bacteria (General Media)
  nonselective for Bacteroides
  nonselective for Campylobacter
  nonselective for Candida
  nonselective for Escherichia coli and Coliforms
  nonselective for Neisseria
  nonselective for Staphylococcus
  nonselective for Streptococcus

Description

Application

Highly nutrient liquid medium for the general purposes, formulated according to USP, FDA and Eur. Phar.regulations.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Campylobacter Detection: Rapid Testing Solutions for the Detection of Campylobacter

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide. Infections are usually caused by consuming cross-contaminated or insufficiently processed food (...
Keywords: Adhesion, Aerobic, Antibiotics, Clinical, Culture media, Gastrointestinal, Gene expression, Homogenization, Microbiology, Polymerase chain reaction

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?