EMAIL THIS PAGE TO A FRIEND
43917 Supelco
SnapIT Ampule Opener
large, for 5-10ml, 10-15ml, 20-25ml ampoules
Purchase
Description
General description
SnapITs are easy-to-use, multi-use ampule openers that avoid the possibility of injury sometimes seen when ampoules are opened by hand.
1. INSERT the ampoule lid into your multi-use SnapIT
2. SNAP open the ampoule
3. EJECT the ampoule lid directly into a sharps container
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?