• USA Home
  • 44600 - Durcupan water-soluble

EMAIL THIS PAGE TO A FRIEND
44600 Sigma-Aldrich

Durcupan water-soluble

set for minimum 10 embeddings

Purchase

Description

Application

water-soluble embedding material for electron microscopy

Legal Information

Durcupan is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 3316 9
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?