• USA Home
  • 45490 - Atrazine-desethyl-2-hydroxy

EMAIL THIS PAGE TO A FRIEND
45490 Supelco

Atrazine-desethyl-2-hydroxy

PESTANAL®, analytical standard

  • CAS Number 19988-24-0

  • Empirical Formula (Hill Notation) C6H11N5O

  • Molecular Weight 169.18

  •  Beilstein/REAXYS Number 2974745

  •  MDL number MFCD00155384

  •  PubChem Substance ID 329756782

  •  NACRES NA.24

Purchase

Properties

Related Categories A-B, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
Quality Level   100
grade   analytical standard
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
storage temp.   room temp
SMILES string   CC(C)Nc1nc(N)nc(O)n1
InChI   1S/C6H11N5O/c1-3(2)8-5-9-4(7)10-6(12)11-5/h3H,1-2H3,(H4,7,8,9,10,11,12)
InChI key   GCKLGRUZDXSATG-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?