• USA Home
  • 462438 - 4-(Diethylamino)benzaldehyde diphenylhydrazone

EMAIL THIS PAGE TO A FRIEND
462438 Sigma-Aldrich

4-(Diethylamino)benzaldehyde diphenylhydrazone

97%

Purchase

Properties

assay   97%
mp   92-94 °C (lit.)
SMILES string   CCN(CC)c1ccc(cc1)\C=N\N(c2ccccc2)c3ccccc3
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
CU5612900

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?