EMAIL THIS PAGE TO A FRIEND
46320 Supelco

Hexestrol

VETRANAL®, analytical standard

Synonym: 4,4′-(1,2-Diethylethylene)diphenol, meso-3,4-Bis(4-hydroxyphenyl)hexane, Dihydrodiethylstilbestrol

  • CAS Number 84-16-2

  • Empirical Formula (Hill Notation) C18H22O2

  • Molecular Weight 270.37

  •  Beilstein/REAXYS Number 3209460

  •  EC Number 201-518-1

  •  MDL number MFCD00068996

  •  PubChem Substance ID 329757136

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Biomarkers and Metabolomics Standards,
grade   analytical standard
product line   VETRANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat
SMILES string   CC[C@H]([C@H](CC)c1ccc(O)cc1)c2ccc(O)cc2
InChI   1S/C18H22O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,17-20H,3-4H2,1-2H3/t17-,18+
InChI key   PBBGSZCBWVPOOL-HDICACEKSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
RTECS 
SL0560850
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?