EMAIL THIS PAGE TO A FRIEND
46866 Supelco

Sulfamoxole

VETRANAL, analytical standard

Synonym: 4-Amino-N-(4,5-dimethyl-2-oxazolyl)benzenesulfonamide, N1-(4,5-Dimethyloxazol-2-yl)sulfanilamide

  • CAS Number 729-99-7

  • Empirical Formula (Hill Notation) C11H13N3O3S

  • Molecular Weight 267.30

  •  Beilstein/REAXYS Number 248434

  •  EC Number 211-982-7

  •  MDL number MFCD00005303

  •  PubChem Substance ID 329757254

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antibiotics Standards, Bacteriostatic,
Quality Level   200
grade   analytical standard
InChI Key   CYFLXLSBHQBMFT-UHFFFAOYSA-N
product line   VETRANAL
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat

Description

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
WO9150000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?