• USA Home
  • 476935 - 1,3,5-Tris(4-hydroxyphenyl)benzene

EMAIL THIS PAGE TO A FRIEND
476935 Sigma-Aldrich

1,3,5-Tris(4-hydroxyphenyl)benzene

97%

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes, T-U-V More...
InChI Key   RQTDWDATSAVLOR-UHFFFAOYSA-N
assay   97%
mp   233-240 °C(lit.)
Gene Information   human ... ESR1(2099)

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?