EMAIL THIS PAGE TO A FRIEND
49335 Sigma-Aldrich

D-Glucuronic acid

purum, ≥97.0% (T)

Synonym: Glucosiduronic acid

  • CAS Number 6556-12-3

  • Empirical Formula (Hill Notation) C6H10O7

  • Molecular Weight 194.14

  •  Beilstein Registry Number 1727083

  •  EC Number 229-486-4

  •  MDL number MFCD00077778

  •  PubChem Substance ID 329757412

Purchase

Properties

Related Categories Acacia, Carbohydrate Chemistry, Carbohydrate Derivatives, Cell Biology, Chemical Biology,
InChI Key   AEMOLEFTQBMNLQ-QIUUJYRFSA-N
grade   purum
assay   ≥97.0% (T)
optical activity   [α]20/D +36.0±2°, 2 hr, c = 6% in H2O
mp   157-161 °C
  159-161 °C(lit.)

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LZ8836600

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G5269 D-Glucuronic acid, ≥98% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?