EMAIL THIS PAGE TO A FRIEND
49597 Supelco

5-Methylquinoxaline

analytical standard

Purchase

Properties

Related Categories Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Almond, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   CQLOYHZZZCWHSG-UHFFFAOYSA-N
assay   ≥98.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.618-1.622
  n20/D 1.62(lit.)
bp   120 °C/15 mmHg(lit.)
mp   20-21 °C (lit.)
density   1.125 g/mL at 25 °C
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
format   neat

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?